missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Ceric Ammonium Sulfate, 0.0200 N (N/50) in 1 N Sulfuric Acid, Ricca Chemical
Chemical Identifiers
CAS |
10378-47-9 |
Molecular Formula |
CeH20N4O18S4 |
Molecular Weight (g/mol) |
632.53 |
InChI Key |
VCNAMBGKEDPVGQ-UHFFFAOYSA-J |
Synonym |
Ceric ammonium sulfate |
PubChem CID |
16211593 |
IUPAC Name |
λâ´-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate |
SMILES |
N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
10378-47-9 |
|
632.53 |
|
Ceric ammonium sulfate |
|
λâ´-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate |
|
CeH20N4O18S4 |
|
VCNAMBGKEDPVGQ-UHFFFAOYSA-J |
|
16211593 |
|
N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O |
Specifications
|
10378-47-9,7732-18-5,7664-93-9 |
|
1.21,96.0,5.27 |
|
Ceric ammonium sulfate |
|
N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O |
|
632.53 |
|
Natural Poly Bottle |
|
120 mL |
|
Ceric Ammonium Sulfate |
|
1.16,92.0,5.07 |
|
CeH20N4O18S4 |
|
VCNAMBGKEDPVGQ-UHFFFAOYSA-J |
|
λâ´-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate |
|
16211593 |
|
1 Normal Sulfuric Acid |
|
0.02Normal |
SDS
missing translation for 'documents'