Learn More
CERILLIANT™ (1R,2S)-(-)-N--Methylephedrine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  Merck Emd Millipore M1291ML
Catalog No. 01001623
Only null left
Explore available promotions
Description
Description
Methylephedrine, a sympathomimetic amine and a bronchodilator, is a component of several over-the-counter cough medications such as 'BRON.' Methylephedrine occasionally used in athletics as a performance enhancing drug for its stimulant effect. This Certified Snap-N-Spike™ Solution is applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from sports testing and urine drug testing to forensic analysis and clinical toxicology.
Specifications
Specifications
67-56-1,552-79-4
MFCD00004504
1.0 mg/mL in methanol
179.26
1 mL
C11H17NO
UN 1230 - class 3 - PG 2 - Methanol, solution
Ephedra
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
CERILLIANT is a registered trademark of Cerilliant Corporation