missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Ethyl-beta-D-glucuronide, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore E0161ML
Description
Ethyl β-D-glucuronide is a primary urinary metabolite of alcohol and can be used as a biomarker for the monitoring of alcohol consumption. This Snap-N-Spike™ Reference Solution is applicable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug testing.Specifications
| Ampule | |
| 9.7°C (49.5°F) - closed cup | |
| C8H14O7 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 222.19 |
| 67-56-1 | |
| 100 μg/mL in methanol | |
| MFCD09037574 | |
| 1 mL |
CERILLIANT is a registered trademark of Cerilliant Corporation