Learn More
CERILLIANT™ Mephedrone-D3 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  Merck Emd Millipore M1391ML
Catalog No. 01001629
Only null left
Explore available promotions
Description
Description
A new internal standard suitable for the quantitation of mephedrone in biological samples using GCMS or LCMS. Mephedrone, also known as 4-methylmethcathinone, is a synthetic stimulant of the amphetamine and cathinone classes. Significant abuse of this new recreational drug has been reported within the last year in the UK; Sweden; and Toronto, Canada with documented seizures in Oregon, Illinois, and Alabama.
Specifications
Specifications
Ampule
9.7°C (49.5°F) - closed cup
C11D3H13ClNO
1 mL
67-56-1
100 μg/mL in methanol (as free base)
UN 1230 - class 3 - PG 2 - Methanol, solution
216.72
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
CERILLIANT is a registered trademark of Cerilliant Corporation