missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cerium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals

$405.41 - $1084.75

Chemical Identifiers

CAS 7440-45-1
Molecular Formula Ce
Molecular Weight (g/mol) 140.12
MDL Number MFCD00010924
InChI Key GWXLDORMOJMVQZ-UHFFFAOYSA-N
Synonym cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
PubChem CID 23974
ChEBI CHEBI:33369
IUPAC Name cerium
SMILES [Ce]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA10141FF
View Documents
Thermo Scientific Chemicals
010141FF
25 x 25 mm
Each for $405.41
 
AA10141FI
View Documents
Thermo Scientific Chemicals
010141FI
50 x 50 mm
Each for $1,084.75
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

7440-45-1
140.12
GWXLDORMOJMVQZ-UHFFFAOYSA-N
23974
cerium
Ce
MFCD00010924
cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
CHEBI:33369
[Ce]

Specifications

Silver to Gray
Ce
UN1333
cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
[Ce]
140.12
CHEBI:33369
Odorless
≈0.42g/25x25mm
Cerium foil, 0.1mm (0.004 in.) thick
7440-45-1
MFCD00010924
25 x 25 mm
GWXLDORMOJMVQZ-UHFFFAOYSA-N
cerium
23974
99.9%
Mylar under Argon
Foil
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: CERIUM

EINECSNumber : 231-154-9

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only