Learn More
Cerium(III) 2,4-pentanedionate hydrate, Thermo Scientific Chemicals
$346.60 - $1565.86
Identifiants chimiques
CAS | 206996-61-4 |
---|---|
Molecular Formula | C15H21CeO6 |
Molecular Weight (g/mol) | 437.44 |
MDL Number | MFCD00150164 |
InChI Key | LCRQZRNVLFDPAZ-UHFFFAOYSA-N |
Synonym | cerium iii acetylacetonate hydrate, cerium iii 2,4-pentanedionate, cerium iii 2,4-pentanedionate hydrate, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy cerio oxy pent-3-en-2-one hydrate |
PubChem CID | 16212434 |
SMILES | [Ce+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
Description
Cerium(III) 2,4-pentanedionate hydrate used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
206996-61-4 | |
437.44 | |
LCRQZRNVLFDPAZ-UHFFFAOYSA-N | |
16212434 |
C15H21CeO6 | |
MFCD00150164 | |
cerium iii acetylacetonate hydrate, cerium iii 2,4-pentanedionate, cerium iii 2,4-pentanedionate hydrate, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy cerio oxy pent-3-en-2-one hydrate | |
[Ce+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
Spécifications
131°C to 132°C | |
206996-61-4 | |
100 g | |
MFCD00150164 | |
Insoluble in water. | |
[Ce+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O | |
437.44 | |
437.44 (Anhydrous) | |
Cerium(III) 2,4-pentanedionate hydrate |
Red to Yellow | |
Powder | |
C15H21CeO6 | |
cerium iii acetylacetonate hydrate, cerium iii 2,4-pentanedionate, cerium iii 2,4-pentanedionate hydrate, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy cerio oxy pent-3-en-2-one hydrate | |
LCRQZRNVLFDPAZ-UHFFFAOYSA-N | |
cerium;(Z)-4-hydroxypent-3-en-2-one;hydrate | |
16212434 | |
Hygroscopic |
Sécurité et manipulation
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 239-720-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only