Learn More
Cerium(III) ammonium nitrate tetrahydrate, Reagent Grade, Thermo Scientific Chemicals
$91.75 - $291.60
Chemical Identifiers
CAS | 13083-04-0 |
---|---|
Molecular Formula | CeH16N7O19 |
Molecular Weight (g/mol) | 558.27 |
MDL Number | MFCD00213483 |
InChI Key | AIVZRMAXALRCHB-UHFFFAOYSA-N |
Synonym | ce.3no3.2hno3.4h2o.2nh3, ammonium cerium iii nitrate tetrahydrate, cerous ammonium nitrate, hydrated, reagent, cerium iii ammonium nitrate tetrahydrate, reagent grade, cerium 3+ bis nitric acid diamine tetrahydrate trinitrate |
PubChem CID | 91886599 |
IUPAC Name | azane;cerium(3+);nitric acid;trinitrate;tetrahydrate |
SMILES | N.N.O.O.O.O.[Ce+3].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Description
A useful oxidant for organic synthesis. This chemical is also useful for the synthesis of quinoxaline derivatives which are useful as DNA cleaving agents, dyes, and organic semiconductors. Used to release organic ligands from metal carbonyls. Whereby the metal is oxidized, Carbon monoxide is produced, and the organic ligand is released.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13083-04-0 | |
558.27 | |
AIVZRMAXALRCHB-UHFFFAOYSA-N | |
91886599 | |
N.N.O.O.O.O.[Ce+3].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
CeH16N7O19 | |
MFCD00213483 | |
ce.3no3.2hno3.4h2o.2nh3, ammonium cerium iii nitrate tetrahydrate, cerous ammonium nitrate, hydrated, reagent, cerium iii ammonium nitrate tetrahydrate, reagent grade, cerium 3+ bis nitric acid diamine tetrahydrate trinitrate | |
azane;cerium(3+);nitric acid;trinitrate;tetrahydrate |
Specifications
White | |
Crystalline | |
CeH16N7O19 | |
ce.3no3.2hno3.4h2o.2nh3, ammonium cerium iii nitrate tetrahydrate, cerous ammonium nitrate, hydrated, reagent, cerium iii ammonium nitrate tetrahydrate, reagent grade, cerium 3+ bis nitric acid diamine tetrahydrate trinitrate | |
AIVZRMAXALRCHB-UHFFFAOYSA-N | |
azane;cerium(3+);nitric acid;trinitrate;tetrahydrate | |
91886599 | |
Reagent | |
Cerium(III) ammonium nitrate tetrahydrate, For reagent grade |
13083-04-0 | |
100 g | |
MFCD00213483 | |
Soluble in water. | |
N.N.O.O.O.O.[Ce+3].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
558.27 | |
558.28 (486.22 Anhydrous) | |
White crystalline |
Safety and Handling
P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H272-H315-H319
DOTInformation : DOT Class: 5.1, Packaging Group: III
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only