missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cerium(III) nitrate hexahydrate, 99.5%, Thermo Scientific Chemicals

$148.01 - $1707.98

Chemical Identifiers

CAS 10294-41-4
Molecular Formula CeH12N3O15
Molecular Weight (g/mol) 434.22
MDL Number MFCD00149631
InChI Key QQZMWMKOWKGPQY-UHFFFAOYSA-N
Synonym cerium iii nitrate hexahydrate, cerium nitrate hexahydrate, cerous nitrate hexahydrate, unii-fl1r38jebb, fl1r38jebb, nitric acid cerium salt, cerium iii nitratehexahydrate, ce.3no3.6h2o, ksc494m5f
PubChem CID 16211466
SMILES O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC218691000
SDS View Documents
Thermo Scientific Chemicals
218691000
100 g Plastic bottle
Each for $148.01
 
AC218690025
SDS View Documents
Thermo Scientific Chemicals
218690025
2.5 kg Plastic bottle
Each for $1,707.98
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10294-41-4
434.22
QQZMWMKOWKGPQY-UHFFFAOYSA-N
16211466
CeH12N3O15
MFCD00149631
cerium iii nitrate hexahydrate, cerium nitrate hexahydrate, cerous nitrate hexahydrate, unii-fl1r38jebb, fl1r38jebb, nitric acid cerium salt, cerium iii nitratehexahydrate, ce.3no3.6h2o, ksc494m5f
O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

10294-41-4
Crystalline Powder or Agglomerating Crystals
99.4% min. (CeO2/TREO basis)
Ce(NO3)3·6H2O
16,67
cerium iii nitrate hexahydrate, cerium nitrate hexahydrate, cerous nitrate hexahydrate, unii-fl1r38jebb, fl1r38jebb, nitric acid cerium salt, cerium iii nitratehexahydrate, ce.3no3.6h2o, ksc494m5f
QQZMWMKOWKGPQY-UHFFFAOYSA-N
cerium(3+) hexahydrate trinitrate
16211466
99.5%
Cerium(III) nitrate hexahydrate, 99.5%
Colorless to White
100 g
CeH12N3O15
MFCD00149631
10,1959
Solubility in water: 1754g/L (25°C). Other solubilities: soluble in alcohol and acetone
O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
434.22
434.22
Plastic bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May intensify fire; oxidizer.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take any precaution to avoid mixing with combustibles.
In case of major

GHS Signal Word: Danger

RTECSNumber : FK6300000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only