missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cerium(III) oxalate hydrate, 99%, Thermo Scientific Chemicals

$136.05 - $471.15

Chemical Identifiers

CAS 15750-47-7
Molecular Formula C6Ce2O12
Molecular Weight (g/mol) 544.29
MDL Number MFCD00013082
InChI Key ZMZNLKYXLARXFY-UHFFFAOYSA-H
Synonym cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate
PubChem CID 91998046
IUPAC Name cerium;oxalic acid;hydrate
SMILES [Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4022714
View Documents
Thermo Scientific Chemicals
04022714
25 g
Each for $136.05
 
AA4022722
View Documents
Thermo Scientific Chemicals
04022722
100 g
Each for $471.15
 
Description

Description

It is used in ceramics, glass, phosphors and pyrotechnics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15750-47-7
544.29
ZMZNLKYXLARXFY-UHFFFAOYSA-H
91998046
[Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
C6Ce2O12
MFCD00013082
cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate
cerium;oxalic acid;hydrate

Specifications

(decomposition)
Crystalline Powder
C6Ce2O12
UN3288
cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate
ZMZNLKYXLARXFY-UHFFFAOYSA-H
cerium;oxalic acid;hydrate
91998046
99%
Hygroscopic
15750-47-7
25 g
MFCD00013082
14,2001
Insoluble in water; Soluble in hot dilute HCl and H2SO4
[Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
544.29
544.29 (Anhydrous)
Odorless
Cerium(III) oxalate hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only