missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cerium ingot, 99.8% min (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 00006530
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gray | |
Rare Earth Oxides Content | |
Dark Gray ingot | |
Ce | |
250 g | |
GWXLDORMOJMVQZ-UHFFFAOYSA-N | |
cerium | |
23974 | |
Odorless |
Cerium | |
7440-45-1 | |
99.8% (REO) | |
MFCD00010924 | |
cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium | |
[Ce] | |
140.12 | |
CHEBI:33369 | |
Ingot |
Chemical Identifiers
7440-45-1 | |
140.12 | |
GWXLDORMOJMVQZ-UHFFFAOYSA-N | |
23974 | |
cerium |
Ce | |
MFCD00010924 | |
cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium | |
CHEBI:33369 | |
[Ce] |
Safety and Handling
Keep container tightly closed and dry.
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : DOT Class: 4.1, Packaging Group: II
EINECSNumber : 231-154-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only