missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min

CAS: 10378-47-9 | (NH4)4Ce(SO4)4·2H2O

$187.12 - $2215.45

Chemical Identifiers

CAS 10378-47-9
Molecular Formula (NH4)4Ce(SO4)4·2H2O
MDL Number MFCD00149417
Synonym Ammonium cerium sulfate
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4019430
View Documents
Thermo Scientific Chemicals
04019430
250 g
Each for $187.12
Only null left
 
AA40194A1
View Documents
Thermo Scientific Chemicals
040194A1
1 kg
Each for $545.48
Only null left
 
AA40194A7
View Documents
Thermo Scientific Chemicals
040194A7
5 kg
Each for $2,215.45
Only null left
 
Description

Description

Ammonium cerium(IV) sulfate dihydrate is an oxidizing agent in organic synthesis and it is used as an analytical reagent, in thin-layer chromatography for the detection of anti-depressant drugs in human blood and urine samples. It is used to wet etch the Co catalyst and synthesis of carbon Nanotubes. It is used in medical and pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10378-47-9
(NH4)4Ce(SO4)4·2H2O
MFCD00149417
Ammonium cerium sulfate

Specifications

130°C
10378-47-9
250 g
(NH4)4Ce(SO4)4·2H2O
Ammonium cerium sulfate
632.56 (596.53 Anhydrous)
Yellow crystalline
Yellow
Crystalline
94% min.
MFCD00149417
Soluble in water and insoluble in ethanol.
ACS
Cerium(IV) ammonium sulfate dihydrate, For analysis ACS
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335-H500

EINECSNumber : 231-567-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only