Learn More
Cerium(IV) isopropoxide, Ce 37-45%, Thermo Scientific Chemicals
$242.88 - $997.10
Chemical Identifiers
CAS | 63007-83-0 |
---|---|
Molecular Formula | C12H28CeO4 |
Molecular Weight (g/mol) | 376.47 |
MDL Number | MFCD00061483 |
InChI Key | BCBBBOXRROHVFG-UHFFFAOYSA-N |
Synonym | cerium iv isopropoxide, acmc-20alht, tetraisopropoxycerium iv, cerium 4+ tetrapropan-2-olate |
PubChem CID | 14473355 |
IUPAC Name | cerium(4+);propan-2-olate |
SMILES | [Ce+4].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-] |
Description
Cerium(IV) isopropoxide, Ce 37-45% is used to catalyzes diastereoselective pinacol couplings. It forms diketonates ,which hydrolyze to CeO2 particles. CeO2 particles have the potential for ion conduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
63007-83-0 | |
376.47 | |
BCBBBOXRROHVFG-UHFFFAOYSA-N | |
14473355 | |
[Ce+4].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-] |
C12H28CeO4 | |
MFCD00061483 | |
cerium iv isopropoxide, acmc-20alht, tetraisopropoxycerium iv, cerium 4+ tetrapropan-2-olate | |
cerium(4+);propan-2-olate |
Specifications
63007-83-0 | |
Orange-Red to Yellow | |
Ce(OC3H7)4 | |
1 g | |
Air and moisture sensitive | |
Soluble in pyridine | |
[Ce+4].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-] | |
376.47 | |
376.48 | |
Powder |
140°C (decomposition) | |
C12H28CeO4 | |
MFCD00061483 | |
UN2925 | |
cerium iv isopropoxide, acmc-20alht, tetraisopropoxycerium iv, cerium 4+ tetrapropan-2-olate | |
BCBBBOXRROHVFG-UHFFFAOYSA-N | |
cerium(4+);propan-2-olate | |
14473355 | |
Ce 37 to 45% | |
Cerium (IV) isopropoxide |
Safety and Handling
GHS H Statement
H314-H228
Causes severe skin burns and eye damage.
Flammable solid.
P210-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H228-H314-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only