Learn More
Cerium(IV) oxide, polishing compound, 57g (2oz), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 039802LA
Description
Polishing compound for IR crystal windows
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
57 g | |
Particles | |
Odorless | |
1306-38-3 | |
MFCD00010933 | |
O=[Ce]=O | |
172.114 | |
CHEBI:79089 |
APS ≈3 Micron | |
Cerium(IV) oxide | |
Polishing compound | |
CeO2 | |
CETPSERCERDGAM-UHFFFAOYSA-N | |
dioxocerium | |
73963 | |
172.12 |
Chemical Identifiers
1306-38-3 | |
172.114 | |
CETPSERCERDGAM-UHFFFAOYSA-N | |
CHEBI:79089 | |
O=[Ce]=O |
CeO2 | |
MFCD00010933 | |
73963 | |
dioxocerium |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 215-150-4
TSCA : Yes
Recommended Storage : Ambient Temperatures
RUO – Research Use Only