Learn More
Cerium(IV) sulfate tetrahydrate, 98+%
CAS: 10294-42-5 | CeO8S2·4H2O | 404.29 g/mol
$478.87 - $1,817.32
Chemical Identifiers
| CAS | 10294-42-5 |
|---|---|
| Molecular Formula | CeO8S2·4H2O |
| Molecular Weight (g/mol) | 404.29 |
| MDL Number | MFCD00149427 |
| InChI Key | MEXSQFDSPVYJOM-UHFFFAOYSA-J |
| Synonym | cerium iv sulfate tetrahydrate, ceric sulfate tetrahydrate, unii-6sax34b8je, 6sax34b8je, acmc-20ecws, ceric sulphate tetrahydrate, ksc493s0d, aronis24302, sulfuric acid, cerium 4+ salt 2:1 , tetrahydrate, cerium iv sulfate hydrate |
| PubChem CID | 22723557 |
| IUPAC Name | cerium(4+);disulfate;tetrahydrate |
| SMILES | O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+4] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC199131000
|
Thermo Scientific Chemicals
199131000 |
100 g | Plastic bottle |
N/A
|
|
||||
|
AC199135000
|
Thermo Scientific Chemicals
199135000 |
500 g | Plastic bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10294-42-5 | |
| 404.29 | |
| MEXSQFDSPVYJOM-UHFFFAOYSA-J | |
| 22723557 | |
| O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+4] |
| CeO8S2·4H2O | |
| MFCD00149427 | |
| cerium iv sulfate tetrahydrate, ceric sulfate tetrahydrate, unii-6sax34b8je, 6sax34b8je, acmc-20ecws, ceric sulphate tetrahydrate, ksc493s0d, aronis24302, sulfuric acid, cerium 4+ salt 2:1 , tetrahydrate, cerium iv sulfate hydrate | |
| cerium(4+);disulfate;tetrahydrate |
Specifications
| 10294-42-5 | |
| Powder | |
| 98% min. (Iodometry) | |
| Ce(SO4)2·4H2O | |
| 02,65; 15,73 | |
| cerium iv sulfate tetrahydrate, ceric sulfate tetrahydrate, unii-6sax34b8je, 6sax34b8je, acmc-20ecws, ceric sulphate tetrahydrate, ksc493s0d, aronis24302, sulfuric acid, cerium 4+ salt 2:1 , tetrahydrate, cerium iv sulfate hydrate | |
| MEXSQFDSPVYJOM-UHFFFAOYSA-J | |
| cerium(4+);disulfate;tetrahydrate | |
| 22723557 | |
| ≥98% | |
| 1.06 |
| Yellow | |
| 100 g | |
| CeO8S2·4H2O | |
| MFCD00149427 | |
| 14, 1990 | |
| Solubility in water: 38g/L (50°C). Other solubilities: soluble in dilute sulfuric acid,slowly soluble in cold,more readily soluble in,hot mineral acids | |
| O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+4] | |
| 404.29 | |
| 404.29 | |
| Plastic bottle | |
| Cerium(IV) sulfate tetrahydrate, 99+% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
RUO – Research Use Only