Learn More
Cerium(IV) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
$90.64 - $318.19
Chemical Identifiers
CAS | 107792-63-2 |
---|---|
Molecular Formula | C4CeF12O12S4 |
Molecular Weight (g/mol) | 736.37 |
MDL Number | MFCD03844746 |
InChI Key | BYAJTISDHIASIP-UHFFFAOYSA-J |
Synonym | cerium iv trifluoromethanesulfonate, cerium iv triflate, cerium trifluoromethanesulfonate, acmc-20alhh, ceric triflate, cerium iv trifluoromethanesulphonate, cerium iv trifluoromethanesulfonate, ?ce cf3so3 4, trifluoromethanesulfonic acid, cerium iv salt, methanesulfonic acid,1,1,1-trifluoro-,cerium 4+ salt 4:1 |
PubChem CID | 9875257 |
IUPAC Name | cerium(4+);trifluoromethanesulfonate |
SMILES | [Ce+4].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL2005606
|
Thermo Scientific Chemicals
L2005606 |
5 g |
Each for $90.64
|
|
|||||
AAL2005614
|
Thermo Scientific Chemicals
L2005614 |
25 g |
Each for $318.19
|
|
|||||
Description
Cerium(IV) trifluoromethanesulfonate, is used as a strong oxidizing agent, in the oxidation of benzyl alcohols and alkylbenzenes to aryl aldehydes and aryl ketones respectively. It can also be used as a catalyst for high-yield deprotection of tert-butyldimethylsilyl ethers
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCerium(IV) trifluoromethanesulfonate, is used as a strong oxidizing agent, in the oxidation of benzyl alcohols and alkylbenzenes to aryl aldehydes and aryl ketones respectively. It can also be used as a catalyst for high-yield deprotection of tert-butyldimethylsilyl ethers
Solubility
Soluble in water.
Notes
Hygroscopic. Store in a cool, dry place in a tightly closed containers. Stable under recommended storage conditions. Incompatible with oxidizing agents and moisture.
Chemical Identifiers
107792-63-2 | |
736.37 | |
BYAJTISDHIASIP-UHFFFAOYSA-J | |
9875257 | |
[Ce+4].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F |
C4CeF12O12S4 | |
MFCD03844746 | |
cerium iv trifluoromethanesulfonate, cerium iv triflate, cerium trifluoromethanesulfonate, acmc-20alhh, ceric triflate, cerium iv trifluoromethanesulphonate, cerium iv trifluoromethanesulfonate, ?ce cf3so3 4, trifluoromethanesulfonic acid, cerium iv salt, methanesulfonic acid,1,1,1-trifluoro-,cerium 4+ salt 4:1 | |
cerium(4+);trifluoromethanesulfonate |
Specifications
107792-63-2 | |
MFCD03844746 | |
Hygroscopic | |
Soluble in water. | |
[Ce+4].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F | |
736.37 | |
736.39 | |
Cerium(IV) trifluoromethanesulfonate |
C4CeF12O12S4 | |
5 g | |
cerium iv trifluoromethanesulfonate, cerium iv triflate, cerium trifluoromethanesulfonate, acmc-20alhh, ceric triflate, cerium iv trifluoromethanesulphonate, cerium iv trifluoromethanesulfonate, ?ce cf3so3 4, trifluoromethanesulfonic acid, cerium iv salt, methanesulfonic acid,1,1,1-trifluoro-,cerium 4+ salt 4:1 | |
BYAJTISDHIASIP-UHFFFAOYSA-J | |
cerium(4+);trifluoromethanesulfonate | |
9875257 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H500
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only