Learn More
Cerium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals
$273.93 - $1387.89
Chemical Identifiers
CAS | 7440-45-1 |
---|---|
Molecular Formula | Ce |
Molecular Weight (g/mol) | 140.12 |
MDL Number | MFCD00010924 |
InChI Key | GWXLDORMOJMVQZ-UHFFFAOYSA-N |
Synonym | cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium |
PubChem CID | 23974 |
ChEBI | CHEBI:33369 |
IUPAC Name | cerium |
SMILES | [Ce] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA44037HF
|
Thermo Scientific Chemicals
044037HF |
10 mm |
Each for $273.93
|
|
|||||
AA44037DJ
|
Thermo Scientific Chemicals
044037DJ |
50 mm |
Each for $772.48
|
|
|||||
AA44037DM
|
Thermo Scientific Chemicals
044037DM |
100 mm |
Each for $1,387.89
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged in mylar under argon
Chemical Identifiers
7440-45-1 | |
140.12 | |
GWXLDORMOJMVQZ-UHFFFAOYSA-N | |
23974 | |
cerium |
Ce | |
MFCD00010924 | |
cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium | |
CHEBI:33369 | |
[Ce] |
Specifications
Silver | |
Ce | |
UN1333 | |
cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium | |
[Ce] | |
140.12 | |
CHEBI:33369 | |
Odorless | |
≈0.21g/mm | |
Cerium rod, ≈6.35mm (0.25 in.) dia. |
7440-45-1 | |
MFCD00010924 | |
10 mm | |
GWXLDORMOJMVQZ-UHFFFAOYSA-N | |
cerium | |
23974 | |
99.9% | |
Mylar under Argon | |
Cast Rod | |
(metals basis excluding Ta) |
Safety and Handling
GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: CERIUM
EINECSNumber : 231-154-9
TSCA : Yes
storageNote1 :
- Packaged in mylar under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only