Mettler Toledo™ Certified Temperature Kits
Quick and easy temperature testing, for use with Mettler Toledo™ Moisture Analyzers
Supplier:  Mettler Toledo™ 30020851
 View more versions of this product
Catalog No. 01915002
Explore available promotions
Description
Description
  • Quick and easy temperature testing
  • Adjusts instrument at the selected operating temperature to ensure reliable results
  • For use with Mettler Toledo™ Moisture Analyzers
Specifications
Specifications
HX204, HS153, HC103 Moisture Analyzer
Temperature Kit