Learn More
Cesium, 99.95+%, (trace metal basis), Thermo Scientific Chemicals
$349.65 - $349.65
Chemical Identifiers
CAS | 7440-46-2 |
---|---|
Molecular Formula | Cs |
Molecular Weight (g/mol) | 132.91 |
MDL Number | MFCD00134037 |
InChI Key | TVFDJXOCXUVLDH-UHFFFAOYSA-N |
Synonym | caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium |
PubChem CID | 5354618 |
ChEBI | CHEBI:30514 |
SMILES | [Cs] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC219190010
|
Thermo Scientific Chemicals
219190010 |
1 g |
Each for $349.65
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
7440-46-2 | |
132.91 | |
TVFDJXOCXUVLDH-UHFFFAOYSA-N | |
5354618 | |
[Cs] |
Cs | |
MFCD00134037 | |
caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium | |
CHEBI:30514 |
Specifications
Golden | |
7440-46-2 | |
1.8730g/mL | |
Cs | |
1 g | |
caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium | |
[Cs] | |
132.91 | |
CHEBI:30514 | |
≥99.95% | |
Cesium, 99.95+% |
28.5°C | |
705.0°C | |
99.95+% | |
MFCD00134037 | |
15, 2007 | |
TVFDJXOCXUVLDH-UHFFFAOYSA-N | |
cesium | |
5354618 | |
132.91 | |
Trace Metal Basis |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
GHS P Statement:
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 231-155-4
RTECSNumber : FK9225000
TSCA : TSCA
RUO – Research Use Only