Learn More
Cesium, 99.98% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01014606
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver | |
7440-46-2 | |
1.873g/cm3 | |
Cs | |
5 g | |
TVFDJXOCXUVLDH-UHFFFAOYSA-N | |
caesium | |
5354618 | |
132.9054 | |
0.0000011 hPa |
Cesium | |
Silver-colored solid | |
99.98% (Metals basis) | |
MFCD00134037 | |
caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium | |
[Cs] | |
132.91 | |
CHEBI:30514 | |
Odorless |
Chemical Identifiers
7440-46-2 | |
132.91 | |
TVFDJXOCXUVLDH-UHFFFAOYSA-N | |
5354618 | |
[Cs] |
Cs | |
MFCD00134037 | |
caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium | |
CHEBI:30514 |
Safety and Handling
P201-P202-P223-P231+P232-P260-P264b-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
H260-H314-H361
DOTInformation : DOT Class: 4.3, Packaging Group: I
EINECSNumber : 231-155-4
RTECSNumber : FK9225000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only