missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cesium Chloride, 99.99%, (Optical Grade I), MP Biomedicals
Solutions are clear and colorless suitable for density gradient work, nucleic acid and protein purification
Supplier: MP Biomedicals Inc 0216004190
Specifications
Cesium Chloride | |
500 g | |
ClCs | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
[Cl-].[Cs+] | |
168.36 | |
CHEBI:63039 | |
Optical I |
7647-17-8 | |
99.99% | |
MFCD00010955 | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
caesium(1+) chloride | |
24293 | |
168.4 |
Chemical Identifiers
7647-17-8 | |
168.36 | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
24293 | |
caesium(1+) chloride |
ClCs | |
MFCD00010955 | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
CHEBI:63039 | |
[Cl-].[Cs+] |
Safety and Handling
Recommended Storage : Room Temperature