missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium Chloride (White Crystalline Powder/Molecular Biology), Fisher BioReagents

Supplier:  Fisher BioReagents BP210500

 View more versions of this product

Catalog No. BP210-500



Explore available promotions

Description

Description

This Molecular Biology-grade cesium chloride is ideal for UV analysis of nucleic acids during density-gradient centrifugation and for the isolation of plasmid DNA, phage DNA, virus particles, and RNA.
Specifications

Specifications

1290°C
646°C
White
Powder/Solid
DNase-, RNase- and Protease-Free
ClCs
15, 2010
AIYUHDOJVYHVIT-UHFFFAOYSA-M
caesium(1+) chloride
24293
168.36
Molecular Biology
1.367 to 1.371 (at 25°C)
Cesium Chloride
7647-17-8
5 to 9
500 g
≥99.999 %
MFCD00010955
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
[Cl-].[Cs+]
168.36
CHEBI:63039
≥99.999%
Poly Bottle

Chemical Identifiers

7647-17-8
168.36
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
caesium(1+) chloride
ClCs
MFCD00010955
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
CHEBI:63039
[Cl-].[Cs+]
Videos
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS Documents

Documents

Promotions

Promotions