missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cesium hydrogen carbonate, 99.99% (metals basis), Thermo Scientific Chemicals
$186.27 - $186.27
Chemical Identifiers
CAS | 29703-01-3 |
---|---|
Molecular Formula | CHCsO3 |
Molecular Weight (g/mol) | 193.92 |
MDL Number | MFCD00010961 |
InChI Key | ZMCUDHNSHCRDBT-UHFFFAOYSA-M |
Synonym | cesium bicarbonate, cesium hydrogen carbonate, caesium bicarbonate, caesium 1+ ion bicarbonate, cesium hydrogencarbonate, acmc-1cmhe, cho3.cs, caesium 1+ bicarbonate, carbonic acid, cesiumsalt 1:1 |
PubChem CID | 2734983 |
IUPAC Name | caesium(1+) hydrogen carbonate |
SMILES | [Cs+].OC([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1423414
|
Thermo Scientific Chemicals
01423414 |
25 g |
Each for $186.27
|
|
|||||
Description
Cesium hydrogen carbonate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
29703-01-3 | |
193.92 | |
ZMCUDHNSHCRDBT-UHFFFAOYSA-M | |
2734983 | |
[Cs+].OC([O-])=O |
CHCsO3 | |
MFCD00010961 | |
cesium bicarbonate, cesium hydrogen carbonate, caesium bicarbonate, caesium 1+ ion bicarbonate, cesium hydrogencarbonate, acmc-1cmhe, cho3.cs, caesium 1+ bicarbonate, carbonic acid, cesiumsalt 1:1 | |
caesium(1+) hydrogen carbonate |
Specifications
175°C (decomposition) | |
Crystalline Aggregates | |
CHCsO3 | |
cesium bicarbonate, cesium hydrogen carbonate, caesium bicarbonate, caesium 1+ ion bicarbonate, cesium hydrogencarbonate, acmc-1cmhe, cho3.cs, caesium 1+ bicarbonate, carbonic acid, cesiumsalt 1:1 | |
ZMCUDHNSHCRDBT-UHFFFAOYSA-M | |
caesium(1+) hydrogen carbonate | |
2734983 | |
99.99% | |
Cesium hydrogen carbonate |
29703-01-3 | |
25 g | |
MFCD00010961 | |
Soluble in water. | |
[Cs+].OC([O-])=O | |
193.92 | |
193.92 | |
Hygroscopic | |
(metals basis) |
Safety and Handling
EINECSNumber : 249-784-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only