Learn More
Cesium hydroxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
$59.81 - $515.47
Chemical Identifiers
CAS | 12260-45-6 |
---|---|
Molecular Formula | CsHO |
Molecular Weight (g/mol) | 149.91 |
MDL Number | MFCD00010964 |
InChI Key | HUCVOHYBFXVBRW-UHFFFAOYSA-M |
Synonym | cesium hydroxide monohydrate, cesium hydroxide hydrate, cesium hydroxide, monohydrate, cesium hydroxide-hydrate, cesium hydroxide, hydrate, ksc493c5l, caesium 1+ ion hydrate hydroxide, cesium hydroxide,monohydrate 9ci |
PubChem CID | 23679066 |
SMILES | [OH-].[Cs+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1323306
|
Thermo Scientific Chemicals
01323306 |
5 g |
Each for $59.81
|
|
|||||
AA1323314
|
Thermo Scientific Chemicals
01323314 |
25 g |
Each for $165.74
|
|
|||||
AA1323322
|
Thermo Scientific Chemicals
01323322 |
100 g |
Each for $515.47
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
12260-45-6 | |
149.91 | |
HUCVOHYBFXVBRW-UHFFFAOYSA-M | |
23679066 |
CsHO | |
MFCD00010964 | |
cesium hydroxide monohydrate, cesium hydroxide hydrate, cesium hydroxide, monohydrate, cesium hydroxide-hydrate, cesium hydroxide, hydrate, ksc493c5l, caesium 1+ ion hydrate hydroxide, cesium hydroxide,monohydrate 9ci | |
[OH-].[Cs+] |
Specifications
12260-45-6 | |
Crystalline | |
99.9% (Metal basis) | |
MFCD00010964 | |
Soluble in water and ethanol. | |
[OH-].[Cs+] | |
149.91 | |
149.91 (Anhydrous) | |
Hygroscopic | |
Cesium hydroxide hydrate |
White | |
5 g | |
CsHO | |
cesium hydroxide monohydrate, cesium hydroxide hydrate, cesium hydroxide, monohydrate, cesium hydroxide-hydrate, cesium hydroxide, hydrate, ksc493c5l, caesium 1+ ion hydrate hydroxide, cesium hydroxide,monohydrate 9ci | |
HUCVOHYBFXVBRW-UHFFFAOYSA-M | |
caesium(1+) hydroxide | |
23679066 | |
Odorless | |
White crystalline |
Safety and Handling
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : DOT Class: 8, Packaging Group: II
EINECSNumber : 244-344-1
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only