missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium Nitrate, 99%+, MilliporeSigma™

Supplier:  Technidata 1028561000

 View more versions of this product

Catalog No. M1028561000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Cesium nitrate
Solid
99%+
UN 1451
NLSCHDZTHVNDCP-UHFFFAOYSA-N
cesium;nitrate
62674
Odorless
Off-white solid
7789-18-6
1 kg
CsNO3
cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j
[N+](=O)([O-])[O-].[Cs+]
194.909
194.91
Plastic bottle

Chemical Identifiers

7789-18-6
194.909
cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j
cesium;nitrate
CsNO3
NLSCHDZTHVNDCP-UHFFFAOYSA-N
62674
[N+](=O)([O-])[O-].[Cs+]
Videos
Safety and Handling

Safety and Handling

H272: May intensify fire; oxidiser.

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 232-146-8

SDS
Documents

Documents

Promotions

Promotions