Excelta™ Chain Nose Pliers
Chain Nose Pliers with radiused edges to reduce scratching and marring of delicate leads. Made of AISI 440 Stainless Steel.
Supplier:  Excelta™ 2949
 View more versions of this product
Catalog No. 17467642
Description
Description
  • Ergonomically designed, ESD-safe handles
Specifications
Specifications
Chain-Nose Pliers
5 in.
39.38 mm
ESD-safe Soft Foam Cushion
0.9 in.
Fine precision
12.7 cm
Stainless Steel
Serrated/Non-serrated
Large Chain Nose