missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chalcone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 159435000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chalcone | |
94-41-7 | |
Yellow | |
>112°C | |
96% min. (GC) | |
C15H12O | |
MFCD00003082 | |
07, 478 | |
chalcone, trans-chalcone, e-chalcone, benzalacetophenone, chalkone, benzylideneacetophenone, phenyl styryl ketone, cinnamophenone, 2-benzalacetophenone, 2-benzylideneacetophenone | |
DQFBYFPFKXHELB-VAWYXSNFSA-N | |
(E)-1,3-diphenylprop-2-en-1-one | |
637760 | |
208.26 | |
Granular Powder |
97% | |
53.0°C to 58.0°C | |
345.0°C to 348.0°C | |
Authentic | |
Plastic bottle | |
C6H5CH=CHCOC6H5 | |
500 g | |
15, 2036 | |
Solubility in water: insoluble. Other solubilities: freely soluble in ether,chloroform,benzene and,carbon disulfide,slightly soluble in alcohol | |
O=C(\C=C\C1=CC=CC=C1)C1=CC=CC=C1 | |
208.26 | |
CHEBI:48965 | |
97% |
Chemical Identifiers
94-41-7 | |
208.26 | |
DQFBYFPFKXHELB-VAWYXSNFSA-N | |
637760 | |
O=C(\C=C\C1=CC=CC=C1)C1=CC=CC=C1 |
C15H12O | |
MFCD00003082 | |
chalcone, trans-chalcone, e-chalcone, benzalacetophenone, chalkone, benzylideneacetophenone, phenyl styryl ketone, cinnamophenone, 2-benzalacetophenone, 2-benzylideneacetophenone | |
CHEBI:48965 |
Safety and Handling
EINECSNumber : 202-330-2
RTECSNumber : FL6900000
TSCA : TSCA
RUO – Research Use Only