Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Supplier:  Thermo Scientific™ FCR500LE
 View more versions of this product
Catalog No. FCR500LE
Explore available promotions
Description
Description
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6 in. (15cm) chart paper and are available as factory or field installed.
Field-installed ink chart recorder
Warranty
Warranty varies by country, please contact us for details
Specifications
Specifications
Inkless Chart Recorder, Field Installed
Revco RDE, Forma FDE, TDE Series, HDE Series (500 box capacity models only)
Boxes (500)
Chart Recorder
Recorder