Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Supplier:  Thermo Scientific™ FCR567TSX
 View more versions of this product
Catalog No. FCR567TSX
Explore available promotions
Description
Description
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6 in. (15cm) chart paper and are available as factory or field installed.
Features:
  • Seven-day recorder
  • Field or factory installed
Ordering information:
  • Field-installed units must be installed by a qualified professional
Specifications
Specifications
Inkless Chart Recorder, Field Installed
-115°C to +50°C
7 Days
Chart Recorder
TSX Series
Recorder