Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Supplier: Thermo Scientific™ FCR600LE
Description
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6 in. (15cm) chart paper and are available as factory or field installed.
Field-installed ink chart recorderWarranty
Warranty varies by country, please contact us for details
Specifications
Inkless Chart Recorder, Field Installed | |
Revco RDE, Forma FDE, TDE Series, HDE Series (600 box capacity models only) | |
Boxes (600) |
Chart Recorder | |
Recorder |