Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Supplier: Thermo Scientific™ FCRP400LE
Description
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6 in. (15cm) chart paper and are available as factory or field installed.
Field-installed inkless chart recorderWarranty
Warranty varies by country, please contact us for details
Specifications
Ink Chart Recorder, Field Installed | |
Chart Recorder | |
Revco RLE, Forma 89000, TLE Series, HLE Series (400 models only) | |
Recorder |