missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lovibond™ CHECKIT™ Comparator Test Kits: Chlorine-Free/Total
Accurate test kits for routine water analysis
Supplier: Lovibond™ L147020
Includes:
- Aqua Comparator Housing (D55 for L147000)
- Aqua Comparator Disc
- DPD No. 1 Rapid Tablets
- DPD No. 3 Rapid Tablets
- 2 x 13.5mm Sample Cells
- Tablet Crusher
- Instructions
- Carrying case
Description
Accurate test kits for routine water analysis
- Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
Specifications
±5% | |
Aqua comparator™ housing, aqua comparator™ disc, DPD No. 1 rapid tablets, DPD No. 3 rapid tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case | |
0 to 4mg/L |
Routine water analysis | |
100 | |
Chlorine Free and Total Test Kit |