missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lovibond™ CHECKIT™ Comparator Test Kits: Chlorine-Free/Total
Accurate test kits for routine water analysis
$240.54 - $262.23
Specifications
Accuracy | ±5% |
---|---|
For Use With (Application) | Routine water analysis |
No. of Tests | 100 |
Type | Chlorine Free and Total Test Kit |
Catalog Number | Mfr. No. | Range | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Range | Includes | Price | Quantity | ||||
09-053-106
|
Lovibond™
L147020 |
0 to 4mg/L | Aqua comparator™ housing, aqua comparator™ disc, DPD No. 1 rapid tablets, DPD No. 3 rapid tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case |
Each for $240.54
|
|
||||
09-053-105
|
Lovibond™
147040 |
0.1 to 2mg/L | Aqua comparator™ housing, aqua comparator™ disc, DPD No. 1 rapid tablets, DPD No. 3 rapid tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case |
Each for $247.40
|
|
||||
09-053-103
|
Lovibond™
147000 |
0.02 to 0.3mg/L | Aqua comparator™ housing, aqua comparator™ disc D55, DPD No. 1 tablets, DPD No. 3 tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case |
Each for $247.40
|
|
||||
09-053-104
|
Lovibond™
147010 |
0 to 1mg/L | Aqua comparator™ housing, aqua comparator™ disc, DPD No. 1 rapid tablets, DPD No. 3 rapid tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case |
Each for $262.23
|
|
||||
Description
Accurate test kits for routine water analysis
- Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
Specifications
±5% | |
Routine water analysis | |
100 | |
Chlorine Free and Total Test Kit |