Learn More
Hanna Instruments™ Checktemp™ Thermometers DFS Item
Supplier:  Hanna Instruments™ HI98501
Numéro de catalogue. 15-077-4
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Spécifications
Spécifications
50 x 185 x 21 mm
1.8 oz.
-58.0°F to +302°F
approx. 2000 hours of continuous use
±0.2°C (-30°C to 120.0°C)
Penetration probe, protective cap, battery
50 g
-50.0°C to +150.0°C
CR2032 Li-ion
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS