CHEMetrics CHEMets Ammonia Test Kit Refill
Provides results in two minutes or less
Supplier: CHEMetrics R1501
Alert:
Contains mercury. Dispose according to local, state or federal laws.
Description
This product contains 30 self-filling CHEMets ampules to refill kit K-1510 for the analysis of ammonia. R-1501 is compatible with C-1501 and C-1510 comparators.Specifications
Ammonia | |
30 self-filling CHEMets ampoules to refill kit K-1510 | |
0 to 1 ppm and 1 to 10 ppm | |
1 min. |
C-1501 and C-1510 comparators | |
30 | |
Direct nesslerization | |
CHEMets Refill Kit |
Warning! Prop 65