CHEMetrics CHEMets Dissolved Oxygen, Visual Water Testing Kit
Provides results in two minutes or less
Supplier: CHEMetrics K7540
Includes: REMOVE
Description
Provide results in two minutes or less
- Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
- To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
- In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
- Comparators have two-year shelf life.
Specifications
Dissolved Oxygen (DO) | |
30 | |
2 min. |
Plastic case, Refill, Comparator, adhesive mounting clamp, permanent mounting clamp, sampling tube and instructions | |
0 to 40 ppb | |
CHEMets Visual Water Testing Kit |