missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
CHEMetrics CHEMets Dissolved Oxygen, Visual Water Testing Kit
Provides results in two minutes or less
$236.74 - $260.94
Specifications
Analytes | Dissolved Oxygen (DO) |
---|---|
No. of Tests | 30 |
Test Time | 2 min. |
Type | CHEMets Visual Water Testing Kit |
Catalog Number | Mfr. No. | Range | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Range | Includes | Price | Quantity | ||||
13-299-201
|
CHEMetrics
K7501 |
0 to 1 ppm | Plastic case, Refill, Comparator, 25 mL sample cup, adhesive mounting clamp, permanent mounting clamp, sampling tube and instructions |
Each for $236.74
|
|
||||
13-299-200
|
CHEMetrics
K7512 |
1 to 12 ppm | Plastic case, Refill, comparator, 25 mL sample cup and instructions |
Each for $236.74
|
|
||||
13-299-202
|
CHEMetrics
K7540 |
0 to 40 ppb | Plastic case, Refill, Comparator, adhesive mounting clamp, permanent mounting clamp, sampling tube and instructions |
Each for $260.94
|
|
||||
13-299-203
|
CHEMetrics
K7599 |
0 to 100 ppb | Plastic case, Refill, Comparator, adhesive mounting clamp, permanent mounting clamp, sampling tube and instructions |
Each for $260.94
|
|
||||
Description
Provide results in two minutes or less
- Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
- To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
- In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
- Comparators have two-year shelf life.
Specifications
Dissolved Oxygen (DO) | |
30 | |
2 min. | |
CHEMets Visual Water Testing Kit |