CHEMetrics Mercury-Free COD Vials Kits
Contains premeasured reagents for accurate, economical determination of COD
Supplier:  CHEMetrics K7366
 View more versions of this product
Catalog No. 13-299-234
Explore available promotions
Description
Description
  • No need to modify or recalibrate existing equipment or procedures
  • Optical-quality glass for accuracy
  • Available in mercury and mercury-free versions
  • Standard 5/8 in. (16 mm) diameter for compatibility with standard COD reactors and spectrophotometers; can also be used in micro-COD reactors
  • Follow the EPA-approved method for compliance reporting and use potassium dichromate as the oxidizing agent, silver sulfate as the catalyst, and mercury chloride to remove chloride interference
  • Mercury-free version uses the same oxidizing agent and catalyst without the chloride compensator
Specifications
Specifications
Chemical Oxygen Demand (COD)
Premeasured reagents for accurate, economical determination of COD, determination of the oxidizable organic matter content of wastewater
150 vials and instruction book
0 to 1500 ppm
2 hr. digestion
A fresh reagent ampoule blank must be prepared for each series of tests; therefore the number of samples that can be tested with each kit will vary.
Digestor block as well as a photometer or spectrophotometer
149 (except distilled water)
Dichromate reactor digestion
COD Vials Kit