missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Chemistry Lab Starter Set

12 Piece set includes everything needed for a basic chemistry laboratory station.

Supplier:  Eisco™ MTST7

Catalog No. S23713


Only null left
Explore available promotions

Description

Description

  • Includes:
    • Base 25 x 16 cm, 1800 g (S24239)
    • Stainless steel rod 750 mm (S02624)
    • Retort ring of 75 mm dia (12-000-108)
    • Double burette clamp (S07858)
    • Retort ring of 100 mm dia. (12-000-109)
    • Two prong adjustable clamp (S41720)
    • Three prong adjustable clamp (S41721)
    • Tripod stand (S14083)
    • Burner tubing (CH0100E)
    • Wire Gauge 150 mm (S95949)
    • Bosshead (S24267)
    • Burner Bunsen (S12809)
Specifications

Specifications

Metal
25.4 mm
Stands and Rings
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions