Learn More
BD Chemotherapy Sharps Collectors
Wide entry ports design for disposal of bulky chemotherapy waste
$544.93 - $544.93
Specifications
Color Yellow
Latex Free Yes
Sterility Non-sterile
Autoclavable Not Autoclavable
Includes Inner gasket lining, biohazard warning label
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (English) Closure Type Price Quantity  
Catalog Number Mfr. No. Capacity (English) Closure Type Price Quantity  
14-826-118
View Documents
BD
305614
17 gal. Hinged Top
Case of 5 Each for $544.93
Only null left
Description
Description
  • Designed to offer increased leak resistance
  • Cannot be autoclaved
  • Yellow
  • Collectors are marked with biohazard warning label
Specifications
Specifications
Yellow
Non-sterile
Inner gasket lining, biohazard warning label
Polypropylene
Yes
Not Autoclavable
For disposal of trace chemotherapy waste
Chemotherapy Sharps Collector
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'