Learn More
Chitosan, 85% deacetylated, Thermo Scientific Chemicals
Chitosan, CAS # 9012-76-4, is a polymer derived from chitin. It is used in the laboratory research of medical material and biomedicine due to its demonstrated bactericidal, antibacterial, anticancer, and antiviral effects.
$102.26 - $429.56
Chemical Identifiers
CAS | 9012-76-4 |
---|---|
Molecular Formula | C56H103N9O39 |
Molecular Weight (g/mol) | 1526.464 |
MDL Number | MFCD00161512 |
InChI Key | FLASNYPZGWUPSU-SICDJOISSA-N |
Synonym | chitosan, poliglusam, deacetylchitin, chicol, flonac c, flonac n, sea cure plus, kytex h, kytex m, kimitsu chitosan f |
PubChem CID | 71853 |
IUPAC Name | methyl N-[(2S,3R,4R,5S,6R)-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-hydroxy-6-(hy |
SMILES | COC(=O)NC1C(C(C(OC1OC2C(OC(C(C2O)N)OC3C(OC(C(C3O)N)O)CO)CO)CO)OC4C(C(C(C(O4)CO)OC5C(C(C(C(O5)CO)OC6C(C(C(C(O6)CO)OC7C(C(C(C(O7)CO)OC8C(C(C(C(O8)CO)OC9C(C(C(C(O9)CO)O)O)N)O)N)O)N)O)N)O)N)O)N)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6414318
|
Thermo Scientific Chemicals
J6414318 |
50 g |
N/A
|
|
|||||
AAJ6414330
|
Thermo Scientific Chemicals
J6414330 |
250 g |
N/A
|
|
|||||
AAJ6414336
|
Thermo Scientific Chemicals
J6414336 |
500 g |
N/A
|
|
|||||
Description
Chitosan is widely utilized in biomedical applications. It finds applications in plantations as natural seed treatment and a growth enhancer. It serves as a fining agent in winemaking and also prevents spoilage. It is involved in the field of medicine to reduce bleeding and as an antibacterial agent. It is used in self-healing coatings which are applied in industrial applications. It is also employed as a part of a filtration process to remove phosphorus, heavy minerals and oils from water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Chitosan is a naturally derived polymer formed from the deacetylation of chitin
- It is an alkaline polysaccharide with moisturizing and adsorption properties
- Derivatives of chitosan are produced by chemically modifying chitosan-reactive functional groups
Application
- Chitosan is used in the laboratory research of medical material and biomedicine due to its bactericidal, antibacterial, anticancer, and antiviral effects
Chemical Identifiers
9012-76-4 | |
1526.464 | |
FLASNYPZGWUPSU-SICDJOISSA-N | |
71853 | |
COC(=O)NC1C(C(C(OC1OC2C(OC(C(C2O)N)OC3C(OC(C(C3O)N)O)CO)CO)CO)OC4C(C(C(C(O4)CO)OC5C(C(C(C(O5)CO)OC6C(C(C(C(O6)CO)OC7C(C(C(C(O7)CO)OC8C(C(C(C(O8)CO)OC9C(C(C(C(O9)CO)O)O)N)O)N)O)N)O)N)O)N)O)N)O |
C56H103N9O39 | |
MFCD00161512 | |
chitosan, poliglusam, deacetylchitin, chicol, flonac c, flonac n, sea cure plus, kytex h, kytex m, kimitsu chitosan f | |
methyl N-[(2S,3R,4R,5S,6R)-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-hydroxy-6-(hy |
Specifications
White | |
9012-76-4 | |
MFCD00161512 | |
Soluble in acetic acid,adipic acid,formic acid,lactic acid,propionic acid and succinic acid.Insoluble in phosphoric acid,sulfuric acid,neutral and alkaline media. | |
FLASNYPZGWUPSU-SICDJOISSA-N | |
methyl N-[(2S,3R,4R,5S,6R)-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-hydroxy-6-(hy | |
71853 |
Powder | |
C56H103N9O39 | |
chitosan, poliglusam, deacetylchitin, chicol, flonac c, flonac n, sea cure plus, kytex h, kytex m, kimitsu chitosan f | |
50 g | |
COC(=O)NC1C(C(C(OC1OC2C(OC(C(C2O)N)OC3C(OC(C(C3O)N)O)CO)CO)CO)OC4C(C(C(C(O4)CO)OC5C(C(C(C(O5)CO)OC6C(C(C(C(O6)CO)OC7C(C(C(C(O7)CO)OC8C(C(C(C(O8)CO)OC9C(C(C(C(O9)CO)O)O)N)O)N)O)N)O)N)O)N)O)N)O | |
1526.464 | |
Chitosan, 85% deacetylated |
Safety and Handling
RTECSNumber : FM6300000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only