Learn More
Chloramine-T trihydrate, 98%
CAS: 7080-50-4 | C7H13ClNNaO5S | 281.68 g/mol
$74.86 - $744.96
Chemical Identifiers
| CAS | 7080-50-4 |
|---|---|
| Molecular Formula | C7H13ClNNaO5S |
| Molecular Weight (g/mol) | 281.68 |
| MDL Number | MFCD00149066 |
| InChI Key | RNROOTWAJBXJBD-UHFFFAOYSA-M |
| Synonym | benzenesulfonamide, n-chloro-4-methyl-, sodium salt, trihydrate |
| PubChem CID | 122197679 |
| IUPAC Name | N-chloro-4-methylbenzenesulfonamide;sodium;trihydrate |
| SMILES | O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1204430
|
Thermo Scientific Chemicals
A1204430 |
250 g |
Each for $74.86
|
|
|||||
|
AAA120440B
|
Thermo Scientific Chemicals
A120440B |
1000 g |
Each for $206.53
|
|
|||||
|
AAA120440I
|
Thermo Scientific Chemicals
A120440I |
5000 g |
Each for $744.96
|
|
|||||
Description
Chloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).
Solubility
Soluble in water and ethyl alcohol. Insoluble in benzene and ethers.
Notes
Incompatible with strong oxidizing agents, acids and ammonia.
Chemical Identifiers
| 7080-50-4 | |
| 281.68 | |
| RNROOTWAJBXJBD-UHFFFAOYSA-M | |
| 122197679 | |
| O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl |
| C7H13ClNNaO5S | |
| MFCD00149066 | |
| benzenesulfonamide, n-chloro-4-methyl-, sodium salt, trihydrate | |
| N-chloro-4-methylbenzenesulfonamide;sodium;trihydrate |
Specifications
| 167°C to 170°C | |
| 250 g | |
| C7H13ClNNaO5S | |
| UN3263 | |
| 192°C (377°F) | |
| benzenesulfonamide, n-chloro-4-methyl-, sodium salt, trihydrate | |
| RNROOTWAJBXJBD-UHFFFAOYSA-M | |
| N-chloro-4-methylbenzenesulfonamide;sodium;trihydrate | |
| 122197679 | |
| 98% | |
| Chloramine-T Trihydrate |
| 7080-50-4 | |
| 98% | |
| MFCD00149066 | |
| 3924168 | |
| 14,2075 | |
| Soluble in water and ethyl alcohol. Insoluble in benzene and ethers. | |
| O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl | |
| 281.68 | |
| 281.69 (227.67 Anhydrous) | |
| Moisture sensitive |
Safety and Handling
GHS H Statement
H334-H314-H318-H302
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H334-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
EINECSNumber : 204-854-7
RTECSNumber : XT5616800
TSCA : Yes
storageNote1 : Moisture Sensitive
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only