missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chloramphenicol Crystalline MP Biomedicals
$81.90 - $2555.33
Chemical Identifiers
CAS | 56-75-7 |
---|---|
Molecular Formula | C11H12Cl2N2O5 |
Molecular Weight (g/mol) | 323.126 |
InChI Key | WIIZWVCIJKGZOK-RKDXNWHRSA-N |
Synonym | chloramphenicol, chlornitromycin, chloromycetin, levomycetin, chlorocid, halomycetin, levomicetina, alficetyn, fenicol, globenicol |
PubChem CID | 5959 |
ChEBI | CHEBI:17698 |
IUPAC Name | 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide |
SMILES | C1=CC(=CC=C1C(C(CO)NC(=O)C(Cl)Cl)O)[N+](=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN19032105
|
MP Biomedicals Inc
0219032105 |
5 g |
Each for $81.90
|
|
|||||
ICN19032125
|
MP Biomedicals Inc
0219032125 |
25 g |
Each for $125.22
|
|
|||||
ICN19032190
|
MP Biomedicals Inc
0219032190 |
500 g |
Each for $1,440.65
|
|
|||||
ICN19032191
|
MP Biomedicals Inc
0219032191 |
1 kg |
Each for $2,555.33
|
|
|||||
Chemical Identifiers
56-75-7 | |
323.126 | |
chloramphenicol, chlornitromycin, chloromycetin, levomycetin, chlorocid, halomycetin, levomicetina, alficetyn, fenicol, globenicol | |
CHEBI:17698 | |
C1=CC(=CC=C1C(C(CO)NC(=O)C(Cl)Cl)O)[N+](=O)[O-] |
C11H12Cl2N2O5 | |
WIIZWVCIJKGZOK-RKDXNWHRSA-N | |
5959 | |
2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide |
Specifications
56-75-7 | |
C11H12Cl2N2O5 | |
chloramphenicol, chlornitromycin, chloromycetin, levomycetin, chlorocid, halomycetin, levomicetina, alficetyn, fenicol, globenicol | |
C1=CC(=CC=C1C(C(CO)NC(=O)C(Cl)Cl)O)[N+](=O)[O-] | |
323.126 | |
CHEBI:17698 | |
Crystalline |
1.586 g/mL | |
5 g | |
WIIZWVCIJKGZOK-RKDXNWHRSA-N | |
2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide | |
5959 | |
323.15 | |
Chloramphenicol |