missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 461020010

 View more versions of this product

Catalog No. AC461020010

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Chlorhexidine
55-56-1
Authentic
97.5% min. (HPLC)
chlorhexidine, chlorhexidinum, chlorhexidin, nolvasan, rotersept, tubulicid, fimeil, hexadol, soretol, cloresidina
C1=CC(=CC=C1NC(=NC(=NCCCCCCN=C(N)N=C(N)NC2=CC=C(C=C2)Cl)N)N)Cl
505.45
CHEBI:3614
98%
134.0°C to 136.0°C
1 g
Glass Bottle
C22H30Cl2N10
GHXZTYHSJHQHIJ-UHFFFAOYSA-N
(1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine
9552079
505.45

Chemical Identifiers

55-56-1
505.45
chlorhexidine, chlorhexidinum, chlorhexidin, nolvasan, rotersept, tubulicid, fimeil, hexadol, soretol, cloresidina
CHEBI:3614
C1=CC(=CC=C1NC(=NC(=NCCCCCCN=C(N)N=C(N)NC2=CC=C(C=C2)Cl)N)N)Cl
C22H30Cl2N10
GHXZTYHSJHQHIJ-UHFFFAOYSA-N
9552079
(1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes serious eye damage. Very toxic to aquatic life with long lasting effects.

GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 200-238-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only