missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

chloro(1,5-cyclooctadiene)iridium(I) dimer, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 363960050

 View more versions of this product

Catalog No. AC363960050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

chloro(1, 5-cyclooctadiene)iridium(I) dimer
190.0°C
C16H24Cl2Ir2
5 g
Solubility in water: insoluble in water
[Cl-].[Cl-].[Ir+].[Ir+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
671.70
671.68
12112-67-3
Glass bottle
MFCD00012414
chloro 1,5-cyclooctadiene iridium i dimer, bis 1,5-cyclooctadiene diiridium i dichloride, 1z,5z-cycloocta-1,5-diene; iridium; dichloride, iridium, di-.mu.-chlorobis 1,2,5,6-.eta.-1,5-cyclooctadiene di, ir2cl2 cod 2, chloro-1,5-cyclooctadiene iridium i dimer, 1z,5z-cycloocta-1,5-diene; iridium 1+
ZFOUDQNHNLDNLD-MIXQCLKLSA-L
(1Z,5Z)-cycloocta-1,5-diene;iridium;dichloride
6436381

Chemical Identifiers

12112-67-3
671.70
ZFOUDQNHNLDNLD-MIXQCLKLSA-L
6436381
[Cl-].[Cl-].[Ir+].[Ir+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
C16H24Cl2Ir2
MFCD00012414
chloro 1,5-cyclooctadiene iridium i dimer, bis 1,5-cyclooctadiene diiridium i dichloride, 1z,5z-cycloocta-1,5-diene; iridium; dichloride, iridium, di-.mu.-chlorobis 1,2,5,6-.eta.-1,5-cyclooctadiene di, ir2cl2 cod 2, chloro-1,5-cyclooctadiene iridium i dimer, 1z,5z-cycloocta-1,5-diene; iridium 1+
(1Z,5Z)-cycloocta-1,5-diene;iridium;dichloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 235-170-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only