Learn More
Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals
$82.25 - $166.76
Chemical Identifiers
CAS | 97-97-2 |
---|---|
Molecular Formula | C4H9ClO2 |
Molecular Weight (g/mol) | 124.56 |
MDL Number | MFCD00000948 |
InChI Key | CRZJPEIBPQWDGJ-UHFFFAOYSA-N |
Synonym | chloroacetaldehyde dimethyl acetal, dimethylchloroacetal, dimethyl chloroacetal, dimethyl chloracetal, ethane, 2-chloro-1,1-dimethoxy, 2-chloro-1,1-dimethoxy-ethane, chloroacetaldehyde, dimethyl acetal, 2-chloroacetaldehyde, dimethyl acetal, methyl chloroacetal, 1-chloro-2,2-dimethoxyethane |
PubChem CID | 60986 |
IUPAC Name | 2-chloro-1,1-dimethoxyethane |
SMILES | COC(CCl)OC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC108502500
|
Thermo Scientific Chemicals
108502500 |
250 mL | Glass bottle |
Each for $82.25
|
|
||||
AC108500010
|
Thermo Scientific Chemicals
108500010 |
1 L | Glass bottle |
Each for $166.76
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
97-97-2 | |
124.56 | |
CRZJPEIBPQWDGJ-UHFFFAOYSA-N | |
60986 | |
COC(CCl)OC |
C4H9ClO2 | |
MFCD00000948 | |
chloroacetaldehyde dimethyl acetal, dimethylchloroacetal, dimethyl chloroacetal, dimethyl chloracetal, ethane, 2-chloro-1,1-dimethoxy, 2-chloro-1,1-dimethoxy-ethane, chloroacetaldehyde, dimethyl acetal, 2-chloroacetaldehyde, dimethyl acetal, methyl chloroacetal, 1-chloro-2,2-dimethoxyethane | |
2-chloro-1,1-dimethoxyethane |
Specifications
97-97-2 | |
Colorless to Yellow | |
128.0°C to 130.0°C | |
Authentic | |
Glass bottle | |
1.4140 to 1.4160 | |
MFCD00000948 | |
1.09 | |
Solubility in water: 52g/L (20 c). | |
COC(CCl)OC | |
124.56 | |
124.57 | |
Liquid |
-73.0°C | |
1.0900g/mL | |
28°C | |
97% | |
C4H9ClO2 | |
ClCH2CH(OCH3)2 | |
250 mL | |
chloroacetaldehyde dimethyl acetal, dimethylchloroacetal, dimethyl chloroacetal, dimethyl chloracetal, ethane, 2-chloro-1,1-dimethoxy, 2-chloro-1,1-dimethoxy-ethane, chloroacetaldehyde, dimethyl acetal, 2-chloroacetaldehyde, dimethyl acetal, methyl chloroacetal, 1-chloro-2,2-dimethoxyethane | |
CRZJPEIBPQWDGJ-UHFFFAOYSA-N | |
2-chloro-1,1-dimethoxyethane | |
60986 | |
97% | |
Chloroacetaldehyde dimethyl acetal, 97% |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 202-624-0
TSCA : TSCA
RUO – Research Use Only