missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroacetamidine hydrochloride, 96%, Thermo Scientific Chemicals

$176.97 - $582.55

Chemical Identifiers

CAS 10300-69-3
Molecular Formula C2H6Cl2N2
Molecular Weight (g/mol) 128.98
MDL Number MFCD00053013
InChI Key GUPOZVHRTJYZCX-UHFFFAOYSA-N
Synonym chloroacetamidine hydrochloride, 2-chloroacetimidamide hydrochloride, 2-chloroethanimidamide hydrochloride, 2-chloroacetamidine hydrochloride, 2-chloroacetamidine hcl, 2-chloroacetamidine monohydrochloride, 2-chloroacetamidinehydrochloride, ethanimidamide, 2-chloro-, monohydrochloride, chloracetamidine hydrochloride, chloroacetamidinehcl
PubChem CID 2776957
IUPAC Name 2-chloroethanimidamide;hydrochloride
SMILES [H+].[Cl-].NC(=N)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1483506
View Documents
Thermo Scientific Chemicals
A1483506
5 g
Each for $176.97
 
AAA1483514
View Documents
Thermo Scientific Chemicals
A1483514
25 g
Each for $582.55
 
Description

Description

Chloroacetamidine hydrochloride is used as a general pharmacophore in delineating characteristics of the amidinotransferase superfamily.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetamidine hydrochloride is used as a general pharmacophore in delineating characteristics of the amidinotransferase superfamily.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

10300-69-3
128.98
GUPOZVHRTJYZCX-UHFFFAOYSA-N
2776957
[H+].[Cl-].NC(=N)CCl
C2H6Cl2N2
MFCD00053013
chloroacetamidine hydrochloride, 2-chloroacetimidamide hydrochloride, 2-chloroethanimidamide hydrochloride, 2-chloroacetamidine hydrochloride, 2-chloroacetamidine hcl, 2-chloroacetamidine monohydrochloride, 2-chloroacetamidinehydrochloride, ethanimidamide, 2-chloro-, monohydrochloride, chloracetamidine hydrochloride, chloroacetamidinehcl
2-chloroethanimidamide;hydrochloride

Specifications

10300-69-3
C2H6Cl2N2
5 g
3678354
chloroacetamidine hydrochloride, 2-chloroacetimidamide hydrochloride, 2-chloroethanimidamide hydrochloride, 2-chloroacetamidine hydrochloride, 2-chloroacetamidine hcl, 2-chloroacetamidine monohydrochloride, 2-chloroacetamidinehydrochloride, ethanimidamide, 2-chloro-, monohydrochloride, chloracetamidine hydrochloride, chloroacetamidinehcl
GUPOZVHRTJYZCX-UHFFFAOYSA-N
2-chloroethanimidamide;hydrochloride
2776957
96%
97°C to 103°C
MFCD00053013
UN3261
Hygroscopic
Insoluble in water.
[H+].[Cl-].NC(=N)CCl
128.98
129
Chloroacetamidine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c

H314-H317-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 233-675-7

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only