missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetic acid sodium salt, 98%

CAS: 3926-62-3 | C2H2ClNaO2 | 116.48 g/mol

$69.18 - $277.70

Chemical Identifiers

CAS 3926-62-3
Molecular Formula C2H2ClNaO2
Molecular Weight (g/mol) 116.48
MDL Number MFCD00002684
InChI Key FDRCDNZGSXJAFP-UHFFFAOYSA-M
Synonym sodium chloroacetate, chloroacetic acid sodium salt, monoxone, acetic acid, chloro-, sodium salt, dow defoliant, sodium monochloroacetate, smca, sodium monochloracetate, caswell no. 755a, chloroctan sodny czech
PubChem CID 23665759
IUPAC Name sodium;2-chloroacetate
SMILES C(C(=O)[O-])Cl.[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC404415000
SDS View Documents
Thermo Scientific Chemicals
404415000
500 g Glass bottle
Each for $69.18
Only null left
 
AC404410025
SDS View Documents
Thermo Scientific Chemicals
404410025
2.5 kg Glass Bottle
Each for $277.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3926-62-3
116.48
FDRCDNZGSXJAFP-UHFFFAOYSA-M
23665759
C(C(=O)[O-])Cl.[Na+]
C2H2ClNaO2
MFCD00002684
sodium chloroacetate, chloroacetic acid sodium salt, monoxone, acetic acid, chloro-, sodium salt, dow defoliant, sodium monochloroacetate, smca, sodium monochloracetate, caswell no. 755a, chloroctan sodny czech
sodium;2-chloroacetate

Specifications

3926-62-3
White
98%
C2H2ClNaO2
MFCD00002684
15, 2112
Solubility in water: soluble in water: 440g/L (20°C).. Other solubilities: slightly soluble in methanol., insoluble in acetone, benzene, ether, ccl4.
C(C(=O)[O-])Cl.[Na+]
116.48
116.48
Crystalline Powder
199.0°C
Authentic
Glass bottle
CH2ClCOONa
500 g
sodium chloroacetate, chloroacetic acid sodium salt, monoxone, acetic acid, chloro-, sodium salt, dow defoliant, sodium monochloroacetate, smca, sodium monochloracetate, caswell no. 755a, chloroctan sodny czech
FDRCDNZGSXJAFP-UHFFFAOYSA-M
sodium;2-chloroacetate
23665759
98%
Chloroacetic acid sodium salt, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Very toxic to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
If skin irritation occurs: Get medical advice/attention.

GHS Signal Word: Danger

EINECSNumber : 223-498-3

RTECSNumber : AG1400000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only