Learn More
Chloroacetone, 96%, stabilized, Thermo Scientific Chemicals
$40.08 - $252.33
Chemical Identifiers
CAS | 78-95-5 |
---|---|
Molecular Formula | C3H5ClO |
Molecular Weight (g/mol) | 92.52 |
MDL Number | MFCD00000936 |
InChI Key | BULLHNJGPPOUOX-UHFFFAOYSA-N |
Synonym | chloroacetone, chloropropanone, acetonyl chloride, monochloroacetone, tonite, 2-propanone, 1-chloro, chloro-2-propanone, 1-chloroacetone, chloromethyl methyl ketone, 1-chloro-2-propanone |
PubChem CID | 6571 |
ChEBI | CHEBI:47220 |
IUPAC Name | 1-chloropropan-2-one |
SMILES | CC(=O)CCl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC173470250
|
Thermo Scientific Chemicals
173470250 |
25 mL | Glass bottle |
Each for $40.08
|
|
||||
AC173472500
|
Thermo Scientific Chemicals
173472500 |
250 mL | Glass bottle |
Each for $91.45
|
|
||||
AC173470010
|
Thermo Scientific Chemicals
173470010 |
1 L | Glass bottle |
Each for $252.33
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
78-95-5 | |
92.52 | |
BULLHNJGPPOUOX-UHFFFAOYSA-N | |
6571 | |
1-chloropropan-2-one |
C3H5ClO | |
MFCD00000936 | |
chloroacetone, chloropropanone, acetonyl chloride, monochloroacetone, tonite, 2-propanone, 1-chloro, chloro-2-propanone, 1-chloroacetone, chloromethyl methyl ketone, 1-chloro-2-propanone | |
CHEBI:47220 | |
CC(=O)CCl |
Specifications
78-95-5 | |
Brown-Yellow to Yellow | |
119.0°C | |
Authentic | |
Glass bottle | |
1.4310 to 1.4330 | |
MFCD00000936 | |
01, 653 | |
15, 2114 | |
Solubility in water: 124g/L (20°C). Other solubilities: soluble in alcohol, ether and chloroform | |
CC(=O)CCl | |
92.52 | |
6571 | |
CHEBI:47220 | |
96% | |
Chloroacetone, Stabilized |
-44.5°C | |
1.1600g/mL | |
32°C | |
95% min. (GC) | |
C3H5ClO | |
ClCH2COCH3 | |
25 mL | |
1.16 | |
chloroacetone, chloropropanone, acetonyl chloride, monochloroacetone, tonite, 2-propanone, 1-chloro, chloro-2-propanone, 1-chloroacetone, chloromethyl methyl ketone, 1-chloro-2-propanone | |
BULLHNJGPPOUOX-UHFFFAOYSA-N | |
1-chloropropan-2-one | |
0.1% epoxidized soybean oil | |
1.11 mPa.s (25°C) | |
92.52 | |
Liquid |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Fatal if inhaled.
Causes ski
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
GHS Signal Word: Danger
EINECSNumber : 201-161-1
RUO – Research Use Only