Learn More
Chlorobenzene, 99+%, for spectroscopy
CAS: 108-90-7 | C6H5Cl | 112.556 g/mol
$163.89 - $163.89
Chemical Identifiers
| CAS | 108-90-7 |
|---|---|
| Molecular Formula | C6H5Cl |
| Molecular Weight (g/mol) | 112.556 |
| MDL Number | MFCD00000530 |
| InChI Key | MVPPADPHJFYWMZ-UHFFFAOYSA-N |
| Synonym | monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene |
| PubChem CID | 7964 |
| ChEBI | CHEBI:28097 |
| IUPAC Name | chlorobenzene |
| SMILES | C1=CC=C(C=C1)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC222115000
|
Thermo Scientific Chemicals
222115000 |
500 mL | Glass Bottle |
Each for $163.89
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopyChemical Identifiers
| 108-90-7 | |
| 112.556 | |
| MVPPADPHJFYWMZ-UHFFFAOYSA-N | |
| 7964 | |
| chlorobenzene |
| C6H5Cl | |
| MFCD00000530 | |
| monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene | |
| CHEBI:28097 | |
| C1=CC=C(C=C1)Cl |
Specifications
| -45.0°C | |
| 0.01 max. at 360nm, 0.07 max. at 300nm, 0.3 max. at 290nm, 1 max. at 287nm | |
| 131.0°C | |
| 23°C | |
| Liquid | |
| 0.0005% max. | |
| 1.1075 | |
| C6H5Cl | |
| 0.8 mPa.s (20°C) | |
| 15, 2122 | |
| Solubility in water: 0.4g/L (20°C). Other solubilities: miscible with most common organic solvents | |
| C1=CC=C(C=C1)Cl | |
| 112.556 | |
| CHEBI:28097 | |
| ≥99% | |
| Chlorobenzene, 99+% |
| 108-90-7 | |
| 1.1075g/mL | |
| 500 mL | |
| Glass Bottle | |
| 1.5230 to 1.5250 | |
| 99+% | |
| C6H5Cl | |
| MFCD00000530 | |
| 05, 199 | |
| monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene | |
| MVPPADPHJFYWMZ-UHFFFAOYSA-N | |
| chlorobenzene | |
| 7964 | |
| 112.56 | |
| Spectroscopy |
Safety and Handling
GHS P Statement
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
GHS P Statement
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 203-628-5
RTECSNumber : CZ0175000
TSCA : TSCA
RUO – Research Use Only