missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorobis[dicyclohexyl(phenyl)phosphino](o-tolyl)nickel(II), TCI America™
Supplier: TCI America C3325100MG
Specifications
| Chlorobis[dicyclohexyl(phenyl)phosphino](o-tolyl)nickel(II) | |
| 150°C | |
| MFCD27978415 | |
| Jamison Catalyst | |
| Crystalline Powder |
| 1419179-26-2 | |
| C43H61ClNiP2 | |
| 100 mg | |
| 734.05 |
Chemical Identifiers
| 1419179-26-2 | |
| C43H61ClNiP2 | |
| MFCD27978415 | |
| Jamison Catalyst |
Safety and Handling
TSCA : No